Learn More
Bochem™ 18/10 Stainless Steel Double Ended Micro-spatulas
Manufactured from stainless steel. Bochem™ 18/10 Stainless Steel Double Ended Micro-spatulas are multi-purpose.
Brand: Bochem™ 3016
This item is not returnable.
View return policy
Description
- Double endet
- Stainless steel
- Autoclavable
- Straight
Specifications
1 Ea. | |
Straight | |
Multi-purpose spatula for a wide range of uses | |
4 mm | |
Stainless Steel (18/10) | |
100 x 4 mm | |
100 mm |
Autoclavable | |
Double Ended Micro Spatula | |
35 mm | |
4 mm | |
2mm | |
Stainless Steel (18/10) |
Product Content Correction
Product Title
Bochem™ 18/10 Stainless Steel Double Ended Micro-spatulas > Size (L x W): 100 x 4mm
Spot an opportunity for improvement?Share a Content Correction