Learn More
Bochem™ 18/10 Stainless Steel Double-Sided Chemical Spoons
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Double-Sided Chemical Spoons are multi-purpose.
41.85 DKK - 58.65 DKK
Specifications
Quantity | 1 Ea. |
---|---|
Product Type | Chemical Scoop |
Handle Material | Stainless Steel (18/10) |
Material | Stainless Steel (18/10) |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
10120591
|
Bochem™
3460 |
120 mm |
41.85 DKK
Each |
Estimated Shipment: 22-08-2025
Log in to see stock. |
||||||
10109970
|
Bochem™
3464 |
250 mm |
58.65 DKK
Each |
Estimated Shipment: 25-08-2025
Log in to see stock. |
||||||
Description
- Chemical spoon
- Double sided
- 18/10 stainless steel
Specifications
1 Ea. | |
Chemical Scoop | |
Stainless Steel (18/10) | |
Stainless Steel (18/10) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Bochem™ 18/10 Stainless Steel Double-Sided Chemical Spoons