Learn More
Bochem™ 18/10 Stainless Steel Rods
187.00 DKK - 744.00 DKK
Product Code | Brand | Length (Metric) | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | For Use With (Equipment) | Price | Quantity & Availability | ||||||
10146410
|
Bochem™
5124 |
1250 mm | Stand bases | N/A | N/A | N/A | |||||
10204561
|
Bochem™
5182 |
750 mm | Stand bases |
272.00 DKK
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
10294221
|
Bochem™
5170 |
500 mm | Stand bases with thread M10 |
187.00 DKK
Each |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
10034980
|
Bochem™
5114 |
1250 mm | Stand bases with thread M10 | N/A | N/A | N/A | |||||
10112461
|
Bochem™
5171 |
600 mm | Stand bases with thread M10 |
205.00 DKK
Each |
Estimated Shipment: 30-06-2025
Log in to see stock. |
||||||
10060911
|
Bochem™
5172 |
750 mm | Stand bases with thread M10 |
235.00 DKK
Each |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
10412331
|
Bochem™
5118 |
1500 mm | Stand bases with thread M10 | N/A | N/A | N/A | |||||
Specifications
Stainless Steel (18/10) | |
Rod | |
1 Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Bochem™ 18/10 Stainless Steel Rods