missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ 19p13/19q13.12 FISH Probe

Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.

12030.00 DKK - 19810.00 DKK

Specifications

Content And Storage Store at 4°C in the dark
Format Tube
For Use With (Application) Fluorescent In Situ Hybridization (Cell)
Product Type DNA/RNA
Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16013365
View Documents
Abnova™
FG0222.100uL
100 μL
12030.00 DKK
100µL
Please to purchase this item. Need a web account? Register with us today!
16023365
View Documents
Abnova™
FG0222.200uL
200 μL
19810.00 DKK
200µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Store at 4°C in the dark
Fluorescent In Situ Hybridization (Cell)
Liquid
Tube
DNA/RNA
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.