Learn More
Abnova™ 19p13.3/19q13.13 FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand: Abnova™ FG0159.100uL
This item is not returnable.
View return policy
Specifications
Store at 4°C in the dark. | |
Tube | |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
Liquid |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
FISH-Ce | |
100 μL |