missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,9H-Perfluoro-1-nonanol, 97%, Thermo Scientific Chemicals

Product Code. 11459367
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 376-18-1
Molecular Formula C9H4F16O
Molecular Weight (g/mol) 432.104
MDL Number MFCD00039629
InChI Key MSXVQELLSMPBFD-UHFFFAOYSA-N
Synonym 1h,1h,9h-hexadecafluoro-1-nonanol, hexadecafluoro-1-nonanol, 1-nonanol, hexadecafluoro, 1-nonanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluoro, 1-nonanol, 1h,1h,9h-hexadecafluoro, 1h,1h,9h-hexadecafluoro-1-hydroxynonane, 1h,1h,9h-perfluoro-1-nonanol, 1h,1h,9h-hexadecafluorononan-1-ol, omega-h-hexadekafluornonanol-1 german, 1h,1h,9h-hexadecafluorononanol
PubChem CID 9779
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorononan-1-ol
SMILES C(C(C(C(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 55°C to 60°C
Boiling Point 157°C to 158°C (200 mmHg)
Quantity 25 g
Beilstein 1808149
Formula Weight 432.11
Percent Purity 97%
Chemical Name or Material 1H,1H,9H-Perfluoro-1-nonanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'