missing translation for 'onlineSavingsMsg'
Learn More
(1S,3R,5R)-PIM447 dihydrochloride, MedChemExpress Product Code.: 16480188

(1S,3R,5R)-PIM447 dihydrochloride, MedChemExpress

Product Code. 16480188
5 mg, 5mg
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16480188

Brand: Medchem Express HY19322C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (1S,3R,5R)-PIM447 (dihydrochloride) an PIM inhibitor extracted from patent US 20100056576 A1, compound example 72, has IC50 values of 0.095 μM for Pim1, 0.522 μM for Pim2 and 0.369 μM for Pim3.

(1S,3R,5R)-PIM447 (dihydrochloride) an PIM inhibitor extracted from patent US 20100056576 A1, compound example 72, has IC50 values of 0.095 μM for Pim1, 0.522 μM for Pim2 and 0.369 μM for Pim3.

Specifications

Chemical Name or Material (1S,3R,5R)-PIM447 dihydrochloride
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H25Cl2F3N4O
Quantity 5 mg
Synonym (1S,3R,5R)-LGH447 dihydrochloride
Solubility Information DMSO : 33.33 mg/mL (64.92 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (24.35 mM; Need ultrasonic)
SMILES O=C(C1=NC(C2=C(F)C=CC=C2F)=C(F)C=C1)NC3=C([C@@H]4C[C@H](N)C[C@H](C)C4)C=CN=C3.Cl.Cl
Molecular Weight (g/mol) 513.38
Formula Weight 513.38
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
(1S,3R,5R)-PIM447 dihydrochloride, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.