Learn More
2-(3-Pyridyl)thioacetamide, 97%, Thermo Scientific™
1368.52 DKK - 5533.00 DKK
Chemical Identifiers
CAS | 51451-44-6 |
---|---|
Molecular Formula | C7H8N2S |
Molecular Weight (g/mol) | 152.215 |
MDL Number | MFCD01691795 |
InChI Key | VSBVYPLNYOJQEV-UHFFFAOYSA-N |
Synonym | 3-pyridineethanethioamide, 2-pyridin-3-yl ethanethioamide, thio-3-pyridineacetamide, 3-pyridineacetamide, thio, 2-3-pyridyl thioacetamide, 2-pyridin-3-yl-thioacetamide, 3-pyridylthioacetamide, 5-22-02-00319 beilstein handbook reference |
PubChem CID | 3039978 |
IUPAC Name | 2-pyridin-3-ylethanethioamide |
SMILES | C1=CC(=CN=C1)CC(=S)N |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
NotesStore in a dry, cool conditions in a well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
51451-44-6 | |
152.215 | |
VSBVYPLNYOJQEV-UHFFFAOYSA-N | |
3039978 | |
C1=CC(=CN=C1)CC(=S)N |
C7H8N2S | |
MFCD01691795 | |
3-pyridineethanethioamide, 2-pyridin-3-yl ethanethioamide, thio-3-pyridineacetamide, 3-pyridineacetamide, thio, 2-3-pyridyl thioacetamide, 2-pyridin-3-yl-thioacetamide, 3-pyridylthioacetamide, 5-22-02-00319 beilstein handbook reference | |
2-pyridin-3-ylethanethioamide |
Specifications
51451-44-6 | |
MFCD01691795 | |
VSBVYPLNYOJQEV-UHFFFAOYSA-N | |
2-pyridin-3-ylethanethioamide | |
3039978 | |
97% |
C7H8N2S | |
3-pyridineethanethioamide, 2-pyridin-3-yl ethanethioamide, thio-3-pyridineacetamide, 3-pyridineacetamide, thio, 2-3-pyridyl thioacetamide, 2-pyridin-3-yl-thioacetamide, 3-pyridylthioacetamide, 5-22-02-00319 beilstein handbook reference | |
C1=CC(=CN=C1)CC(=S)N | |
152.215 | |
152.21 | |
2-(3-Pyridyl)thioacetamide |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures