missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2,4,6-Trimethylbenzyl mercaptan, 97%
Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Melting Point | 46°C to 47°C |
Boiling Point | 120°C (5 mmHg) |
Odor | Unpleasant |
Quantity | 5 g |
UN Number | UN2811 |
Sensitivity | Air Sensitive |
Formula Weight | 166.29 |
Percent Purity | 97% |
Chemical Name or Material | 2,4,6-Trimethylbenzyl mercaptan |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction