missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3-methylthiophene, 98%, Thermo Scientific Chemicals

288.69 DKK - 2453.00 DKK

Chemical Identifiers

CAS 13191-36-1
Molecular Formula C5H4Br2S
Molecular Weight (g/mol) 255.955
MDL Number MFCD00015470
InChI Key IHFXZROPBCBLLG-UHFFFAOYSA-N
Synonym thiophene, 2,5-dibromo-3-methyl, 2,5-dibromo-3-methyl-thiophene, acmc-1bt3y, 2-p-nitrophenylthio ethanol, 2,5-dibrom-3-methylthiophen, 3-methyl-2,5-dibromothiophene, 2,5-dibromo-3-methylthiophene
PubChem CID 83219
IUPAC Name 2,5-dibromo-3-methylthiophene
SMILES CC1=C(SC(=C1)Br)Br
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11337795
SDS View Documents
Thermo Scientific Alfa Aesar
L02693.06
5 g
288.69 DKK
5g
Please to purchase this item. Need a web account? Register with us today!
11347795
SDS View Documents
Thermo Scientific Alfa Aesar
L02693.14
25 g
914.79 DKK
25g
Please to purchase this item. Need a web account? Register with us today!
11357795
SDS View Documents
Thermo Scientific Alfa Aesar
L02693.22
100 g N/A N/A N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'einecsNumber' : 236-147-4

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Dibromo-3-methylthiophene, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.