Learn More
2,6-Dichloro-4-(trifluoromethyl)aniline, 97%, Thermo Scientific™
178.00 DKK - 551.00 DKK
Chemical Identifiers
CAS | 24279-39-8 |
---|---|
Molecular Formula | C7H4Cl2F3N |
Molecular Weight (g/mol) | 230.01 |
MDL Number | MFCD00052918 |
InChI Key | ITNMAZSPBLRJLU-UHFFFAOYSA-N |
Synonym | 2,6-dichloro-4-trifluoromethyl aniline, 4-amino-3,5-dichlorobenzotrifluoride, 3,5-dichloro-4-amino benzotrifluoride, 3,5-dichloro-4-aminobenzotrifluoride, 2,6-dichloro-4-trifluoromethyl benzenamine, benzenamine, 2,6-dichloro-4-trifluoromethyl, 2,6-dichloro-4-trifluoromethyl phenylamine, 2,6-dichloro-4-trifluoromethyl-benzenamine, pubchem2187Show More |
PubChem CID | 141094 |
IUPAC Name | 2,6-dichloro-4-(trifluoromethyl)aniline |
SMILES | NC1=C(Cl)C=C(C=C1Cl)C(F)(F)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
24279-39-8 | |
230.01 | |
ITNMAZSPBLRJLU-UHFFFAOYSA-N | |
141094 | |
NC1=C(Cl)C=C(C=C1Cl)C(F)(F)F |
C7H4Cl2F3N | |
MFCD00052918 | |
2,6-dichloro-4-trifluoromethyl aniline, 4-amino-3,5-dichlorobenzotrifluoride, 3,5-dichloro-4-amino benzotrifluoride, 3,5-dichloro-4-aminobenzotrifluoride, 2,6-dichloro-4-trifluoromethyl benzenamine, benzenamine, 2,6-dichloro-4-trifluoromethyl, 2,6-dichloro-4-trifluoromethyl phenylamine, 2,6-dichloro-4-trifluoromethyl-benzenamine, pubchem2187Show More | |
2,6-dichloro-4-(trifluoromethyl)aniline |
Specifications
24279-39-8 | |
1.532 | |
88°C (190°F) | |
MFCD00052918 | |
2838819 | |
ITNMAZSPBLRJLU-UHFFFAOYSA-N | |
2,6-dichloro-4-(trifluoromethyl)aniline | |
141094 | |
97% |
34°C to 37°C | |
60°C to 62°C (1 mmHg) | |
C7H4Cl2F3N | |
UN3077 | |
2,6-dichloro-4-trifluoromethyl aniline, 4-amino-3,5-dichlorobenzotrifluoride, 3,5-dichloro-4-amino benzotrifluoride, 3,5-dichloro-4-aminobenzotrifluoride, 2,6-dichloro-4-trifluoromethyl benzenamine, benzenamine, 2,6-dichloro-4-trifluoromethyl, 2,6-dichloro-4-trifluoromethyl phenylamine, 2,6-dichloro-4-trifluoromethyl-benzenamine, pubchem2187 | |
NC1=C(Cl)C=C(C=C1Cl)C(F)(F)F | |
230.01 | |
230.02 | |
2,6-Dichloro-4-(trifluoromethyl)aniline |
Safety and Handling
GHS H Statement
H302-H332-H315-H317
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
H302+H332-H315-H317-H319
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 416-430-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.