missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethoxybenzoic acid, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
It is used in the synthesis of indoles as well as coumarins related to Novobiocin, used as potent inhibitors of heat-shock protein 90 in the treatment of breast cancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the synthesis of indoles as well as coumarins related to Novobiocin, used as potent inhibitors of heat-shock protein 90 in the treatment of breast cancer.
Solubility
Insoluble in water. Solubility in methanol is almost transparent. Soluble in DMSO, Ethyl Acetate and acetone.
Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used in the synthesis of indoles as well as coumarins related to Novobiocin, used as potent inhibitors of heat-shock protein 90 in the treatment of breast cancer.
Solubility
Insoluble in water. Solubility in methanol is almost transparent. Soluble in DMSO, Ethyl Acetate and acetone.
Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
Melting Point | ∼188°C (decomposition) |
Odor | Odorless |
Quantity | 100 g |
Beilstein | 2212190 |
Solubility Information | Insoluble in water. Solubility in methanol is almost transparent. Soluble in DMSO,Ethyl Acetate and acetone. |
Formula Weight | 182.18 |
Percent Purity | 99% |
Chemical Name or Material | 2,6-Dimethoxybenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'