missing translation for 'onlineSavingsMsg'
Learn More
2,6-Diphenylpyridine, 97%, Thermo Scientific Chemicals Product Code.: 11410638

2,6-Diphenylpyridine, 97%, Thermo Scientific Chemicals

Product Code. 11410638
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11410638

Brand: Thermo Scientific Alfa Aesar B25179.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3558-69-8 | C17H13N | 231.298 g/mol

2,6-Diphenylpyridine is used as a tridentate [C?N?C] dianionic ligand in organogold(III) chemistry, an intermediate. It readily adds an electron from alkali metals in THF to give a radical anion which is a powerful reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Diphenylpyridine is used as a tridentate [C∧N∧C] dianionic ligand in organogold(III) chemistry, an intermediate. It readily adds an electron from alkali metals in THF to give a radical anion which is a powerful reducing agent.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3558-69-8
Molecular Formula C17H13N
Molecular Weight (g/mol) 231.298
MDL Number MFCD00006284
InChI Key PJUOHDQXFNPPRF-UHFFFAOYSA-N
Synonym pyridine, 2,6-diphenyl, unii-zi75v15y1t, 2,6-diphenyl-pyridine, 2,6-diphenyl pyridine, maybridge3_000511, acmc-1cq41, pjuohdqxfnpprf-uhfffaoysaShow MoreShow Less
PubChem CID 72920
IUPAC Name 2,6-diphenylpyridine
SMILES C1=CC=C(C=C1)C2=NC(=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 74°C to 81°C
Boiling Point 210°C (3 mmHg)
Quantity 5 g
Beilstein 7516
Solubility Information Insoluble in water.
Formula Weight 231.3
Percent Purity 97%
Chemical Name or Material 2,6-Diphenylpyridine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10229433 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25179.14,B25179.06
Lot Number10234065 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25179.14,B25179.06
Lot Number10196610 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25179.14,B25179.06
Lot Number10222345 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25179.14,B25179.06
Lot Number10149750 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25179.14,B25179.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,6-Diphenylpyridine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.