Learn More
2-([(9H-Fluoren-9-ylmethoxy)carbonyl]amino)-4-methylpentanoic acid, 97%, Thermo Scientific™ Product Code.: 10799595
2-([(9H-Fluoren-9-ylmethoxy)carbonyl]amino)-4-methylpentanoic acid, 97%, Thermo Scientific™
Product Code. 10799595
Amber Glass Bottle; 10g
Click to view available options
:
Amber Glass Bottle; 10g
Product Code. 10799595
Brand: Thermo Scientific Maybridge NRB05198EA
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 35661-60-0
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Molecular Formula C21H23NO4
Linear Formula FMOC-NHCH(COOH)CH2CH(CH3)2
MDL Number MFCD00037133
Quantity 10g
Specific Rotation Condition − 26.00 (24.00°C c=1,DMF)
Synonym fmoc-l-leucine, fmoc-leu-oh, fmoc-l-leu-oh, n-fmoc-l-leucine, n-9-fluorenylmethoxycarbonyl-l-leucine, fmoc-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-l-leucine, unii-kf18f70uf3, fmoc-leu, n-alpha-fmoc-l-leucine
InChI Key CBPJQFCAFFNICX-IBGZPJMESA-N
SMILES CC(C)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoic acid
Molecular Weight (g/mol) 353.418
PubChem CID 1549133
Formula Weight 353.42
Chemical Name or Material 2-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-4-methylpentanoic acid
Show More Show Less
Product Content Correction
Product Title
2-([(9H-Fluoren-9-ylmethoxy)carbonyl]amino)-4-methylpentanoic acid, 97%, Thermo Scientific™ >