missing translation for 'onlineSavingsMsg'
Learn More
2-Amino-2',5-dichlorobenzophenone, 98% Product Code.: 11414267

2-Amino-2',5-dichlorobenzophenone, 98%

Product Code. 11414267
50 g, 50g
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
This item is not returnable. View return policy

Product Code. 11414267

Brand: Thermo Scientific Alfa Aesar B20360.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2958-36-3 | C13H9Cl2NO | 266.121 g/mol

2-Amino-2',5-dichlorobenzophenone is used to produce lorazepam a drug of the benzodiazepine group, used especially to treat anxiety.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-2′,5-dichlorobenzophenone is used to produce lorazepam a drug of the benzodiazepine group, used especially to treat anxiety.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2958-36-3
Molecular Formula C13H9Cl2NO
Molecular Weight (g/mol) 266.121
MDL Number MFCD00007840
InChI Key KWZYIAJRFJVQDO-UHFFFAOYSA-N
Synonym 2-amino-2',5-dichlorobenzophenone, 2-amino-5-chlorophenyl 2-chlorophenyl methanone, 4-chloro-2-2-chlorobenzoyl aniline, methanone, 2-amino-5-chlorophenyl 2-chlorophenyl, unii-g806veo3ke, 2-amino-5-chloro-2'-chlorobenzophenone, 2-amino-5-chlorophenyl-2-chlorophenyl methanone, g806veo3ke, benzophenone, 2-amino-2',5-dichloro, 2-amino-5,2'-dichlorobenzophenone
PubChem CID 18069
IUPAC Name (2-amino-5-chlorophenyl)-(2-chlorophenyl)methanone
SMILES C1=CC=C(C(=C1)C(=O)C2=C(C=CC(=C2)Cl)N)Cl

Specifications

Melting Point 87°C to 89°C
Odor Odorless
Quantity 50 g
Beilstein 652774
Solubility Information Insoluble in water.
Formula Weight 266.13
Percent Purity 99%
Chemical Name or Material 2-Amino-2',5-dichlorobenzophenone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberC6480A Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB20360.06,B20360.18,B20360.09
Lot NumberUSLF006754 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB20360.06,B20360.18,B20360.09
Lot NumberE3050C Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB20360.06,B20360.18,B20360.09
Lot Number10226273 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB20360.06,B20360.18,B20360.09
Lot Number10228792 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB20360.06,B20360.18,B20360.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Amino-2',5-dichlorobenzophenone, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.