Learn More
Thermo Scientific Chemicals 2-Amino-2'-deoxyadenosine, 99% Product Code.: 15476289
Thermo Scientific Chemicals 2-Amino-2'-deoxyadenosine, 99%
Product Code. 15476289
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
Product Code. 15476289
Brand: Thermo Scientific Chemicals J64266.06
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4546-70-7
Molecular Formula C10H14N6O3
Molecular Weight (g/mol) 266.26
MDL Number MFCD00047240
InChI Key NOLHIMIFXOBLFF-DWFKHXNSNA-N
Synonym 2-amino-2'-deoxyadenosine, 2,6-diaminopurine-2'-deoxyriboside, adenosine, 2-amino-2'-deoxy, 2,6-diaminopurinedeoxyriboside, 2,6-diaminopurine 2'-deoxyriboside, unii-4zph2f1jhr, 4zph2f1jhr, 9h-purine, 9-2-deoxy-beta-d-erythro-pentofuranosyl-2,6-diamino, 2-amino-deoxyadenosine, 2-amino-2/'-deoxyadenosine
PubChem CID 97188
IUPAC Name (2R,3S,5R)-5-(2,6-diaminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
SMILES NC1=NC(N)=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=N1
Specifications
Color White
Quantity 5 g
Formula Weight 266.26
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Amino-2'-deoxyadenosine
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS09I018 Certificate TypeCertificate of Analysis Date27/06/2022 Catalog NumberJ64266.03,J64266.14,J64266.06
Lot NumberW07H005 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ64266.03,J64266.14,J64266.06
Lot NumberH31Y036 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ64266.03,J64266.14,J64266.06
Lot NumberH20Z044 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ64266.03,J64266.14,J64266.06
Lot NumberY02G013 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ64266.03,J64266.14,J64266.06
RUO – Research Use Only
Product Content Correction
Product Title
Thermo Scientific Chemicals 2-Amino-2'-deoxyadenosine, 99% >