missing translation for 'onlineSavingsMsg'
Learn More
2-Amino-6-chlorobenzonitrile, 98% Product Code.: 11327036

2-Amino-6-chlorobenzonitrile, 98%

Product Code. 11327036
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 11327036

Brand: Thermo Scientific Alfa Aesar L06883.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

CAS: 6575-11-7 | C7H5ClN2 | 152.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6575-11-7
MDL Number MFCD00015546
PubChem CID 81056
IUPAC Name 2-amino-6-chlorobenzonitrile
SMILES C1=CC(=C(C(=C1)Cl)C#N)N

Specifications

Melting Point 133°C to 137°C
Quantity 5 g
Molecular Formula C7H5ClN2
UN Number UN3439
Beilstein 2717090
Synonym 6-chloroanthranilonitrile, benzonitrile, 2-amino-6-chloro, 2-amino-6-chloro-benzonitrile, 3-chloro-2-cyanoaniline, pubchem4614, acmc-1b6zr, 6-amino-2-chlorobenzonitrile, ksc497g6t, 2-amino-6-chloro benzonitrile, benzonitrile,2-amino-6-chloro
InChI Key MEJVTQKBWPYBFG-UHFFFAOYSA-N
Molecular Weight (g/mol) 152.58
Formula Weight 152.58
Percent Purity 98%
Chemical Name or Material 2-Amino-6-chlorobenzonitrile
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Product Code
Lot Number10160013 Certificate TypeCertificate of Analysis Date30/08/2024 Product CodeL06883.06,L06883.03
Lot Number10160013 Certificate TypeCertificate of Analysis Date06/12/2021 Product CodeL06883.06,L06883.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Amino-6-chlorobenzonitrile, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.