missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3,5-trimethyl-4-nitrobenzene, 90+%, Thermo Scientific Chemicals

508.38 DKK - 1829.57 DKK

Chemical Identifiers

CAS 90561-85-6
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.088
MDL Number MFCD00015924
InChI Key IUFDRRDWXUSFBG-UHFFFAOYSA-N
Synonym 3-bromo-2,4,6-trimethylnitrobenzene, 2-bromo-4-nitro-1,3,5-trimethylbenzene, 1-bromo-3-nitro-2,4,6-trimethylbenzene, 4-bromo-2-nitromesitylene, 4-bromo-1,3,5-trimethyl-2-nitrobenzene, 2-bromanyl-1,3,5-trimethyl-4-nitro-benzene
PubChem CID 2757200
IUPAC Name 2-bromo-1,3,5-trimethyl-4-nitrobenzene
SMILES CC1=CC(=C(C(=C1[N+](=O)[O-])C)Br)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11391827
View Documents
Thermo Scientific Alfa Aesar
L05107.03
1 g
508.38 DKK
1g
Please to purchase this item. Need a web account? Register with us today!
11301837
View Documents
Thermo Scientific Alfa Aesar
L05107.06
5 g
1829.57 DKK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

90561-85-6
244.088
IUFDRRDWXUSFBG-UHFFFAOYSA-N
2757200
CC1=CC(=C(C(=C1[N+](=O)[O-])C)Br)C
C9H10BrNO2
MFCD00015924
3-bromo-2,4,6-trimethylnitrobenzene, 2-bromo-4-nitro-1,3,5-trimethylbenzene, 1-bromo-3-nitro-2,4,6-trimethylbenzene, 4-bromo-2-nitromesitylene, 4-bromo-1,3,5-trimethyl-2-nitrobenzene, 2-bromanyl-1,3,5-trimethyl-4-nitro-benzene
2-bromo-1,3,5-trimethyl-4-nitrobenzene

Specifications

90561-85-6
106°C to 107°C (1.3 mmHg)
MFCD00015924
Light sensitive
IUFDRRDWXUSFBG-UHFFFAOYSA-N
2-bromo-1,3,5-trimethyl-4-nitrobenzene
2757200
≥90%
56°C to 62°C
C9H10BrNO2
1958703
3-bromo-2,4,6-trimethylnitrobenzene, 2-bromo-4-nitro-1,3,5-trimethylbenzene, 1-bromo-3-nitro-2,4,6-trimethylbenzene, 4-bromo-2-nitromesitylene, 4-bromo-1,3,5-trimethyl-2-nitrobenzene, 2-bromanyl-1,3,5-trimethyl-4-nitro-benzene
CC1=CC(=C(C(=C1[N+](=O)[O-])C)Br)C
244.088
244.09
2-Bromo-1,3,5-trimethyl-4-nitrobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.