missing translation for 'onlineSavingsMsg'
Learn More

2-bromo-5-fluorophenyl isothiocyanate, Thermo Scientific™

84.31 DKK - 432.74 DKK

Chemical Identifiers

CAS 175205-35-3
Molecular Formula C7H3BrFNS
Molecular Weight (g/mol) 232.07
MDL Number MFCD00673194
InChI Key SSLVXFSUADEZBQ-UHFFFAOYSA-N
Synonym 2-bromo-4-fluorophenyl isothiocyanate, 2-bromo-4-fluorophenylisothiocyanate, pubchem5018, acmc-20ao2g, 2-bromo-4-fluorobenzenisothiocyanate, benzene,1-bromo-4-fluoro-2-isothiocyanato, 2-bromo-4-fluorophenyl isothiocyanate, tech., benzene, 2-bromo-4-fluoro-1-isothiocyanato, 2-bromanyl-4-fluoranyl-1-isothiocyanato-benzene, benzene, 2-bromo-4-fluoro-1-isothiocyanato-9ci
PubChem CID 2736332
IUPAC Name 2-bromo-4-fluoro-1-isothiocyanatobenzene
SMILES FC1=CC=C(N=C=S)C(Br)=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10205402
View Documents
Thermo Scientific Maybridge
SB01713DA
1 g
84.31 DKK
1g
Please to purchase this item. Need a web account? Register with us today!
10312315
View Documents
Thermo Scientific Maybridge
SB01713DE
5 g
264.12 DKK
5g
Please to purchase this item. Need a web account? Register with us today!
10260983
View Documents
Thermo Scientific Maybridge
SB01713EA
10 g
432.74 DKK
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

175205-35-3
232.07
SSLVXFSUADEZBQ-UHFFFAOYSA-N
2736332
FC1=CC=C(N=C=S)C(Br)=C1
C7H3BrFNS
MFCD00673194
2-bromo-4-fluorophenyl isothiocyanate, 2-bromo-4-fluorophenylisothiocyanate, pubchem5018, acmc-20ao2g, 2-bromo-4-fluorobenzenisothiocyanate, benzene,1-bromo-4-fluoro-2-isothiocyanato, 2-bromo-4-fluorophenyl isothiocyanate, tech., benzene, 2-bromo-4-fluoro-1-isothiocyanato, 2-bromanyl-4-fluoranyl-1-isothiocyanato-benzene, benzene, 2-bromo-4-fluoro-1-isothiocyanato-9ci
2-bromo-4-fluoro-1-isothiocyanatobenzene

Specifications

175205-35-3
MFCD00673194
SSLVXFSUADEZBQ-UHFFFAOYSA-N
2-bromo-4-fluoro-1-isothiocyanatobenzene
2736332
Tech.
2-Bromo-5-fluorophenyl isothiocyanate
C7H3BrFNS
2-bromo-4-fluorophenyl isothiocyanate, 2-bromo-4-fluorophenylisothiocyanate, pubchem5018, acmc-20ao2g, 2-bromo-4-fluorobenzenisothiocyanate, benzene,1-bromo-4-fluoro-2-isothiocyanato, 2-bromo-4-fluorophenyl isothiocyanate, tech., benzene, 2-bromo-4-fluoro-1-isothiocyanato, 2-bromanyl-4-fluoranyl-1-isothiocyanato-benzene, benzene, 2-bromo-4-fluoro-1-isothiocyanato-9ci
FC1=CC=C(N=C=S)C(Br)=C1
232.07
232.08
Technical
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications