Learn More
2-Bromo-6-(4-morpholinyl)benzonitrile, 98%, Thermo Scientific Chemicals Product Code.: 15487698
2-Bromo-6-(4-morpholinyl)benzonitrile, 98%, Thermo Scientific Chemicals
Product Code. 15487698
1 g
Click to view available options
Quantity:
250 mg
1 g
Product Code. 15487698
Brand: Thermo Scientific Alfa Aesar H61193.03
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1129540-65-3
Molecular Formula C11H11BrN2O
Molecular Weight (g/mol) 267.13
MDL Number MFCD11037772
InChI Key HKIGGSMIIBGCKZ-UHFFFAOYSA-N
Synonym 2-bromo-6-morpholinobenzonitrile, 2-bromo-6-morpholin-4-yl benzonitrile
PubChem CID 59588248
SMILES BrC1=CC=CC(N2CCOCC2)=C1C#N
Specifications
Quantity 1 g
UN Number UN3439
IUPAC Name 2-bromo-6-(morpholin-4-yl)benzonitrile
Formula Weight 267.13
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-6-(4-morpholinyl)benzonitrile
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM11E040 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61193.03,H61193.MD
RUO – Research Use Only
Product Content Correction
Product Title
2-Bromo-6-(4-morpholinyl)benzonitrile, 98%, Thermo Scientific Chemicals >