missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylaniline, 98%, Thermo Scientific Chemicals

Product Code. 11367765
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-63-8
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD00007679
InChI Key WFNLHDJJZSJARK-UHFFFAOYSA-N
Synonym 6-chloro-2-methylaniline, 6-chloro-o-toluidine, 2-amino-3-chlorotoluene, benzenamine, 2-chloro-6-methyl, 3-chloro-2-aminotoluene, 6-chloro-2-toluidine, 2-methyl-6-chloroaniline, o-toluidine, 6-chloro, 2-chloro-6-methyl-aniline, 2-chloro-6-methylaninile
PubChem CID 6897
IUPAC Name 2-chloro-6-methylaniline
SMILES CC1=CC=CC(Cl)=C1N

Specifications

Melting Point 2°C
Density 1.174
Boiling Point 215°C
Flash Point 98°C (208°F)
Refractive Index 1.5775
Quantity 1 g
UN Number UN3429
Beilstein 774624
Formula Weight 141.6
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-methylaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'