missing translation for 'onlineSavingsMsg'
Learn More
2-Fluorobenzothiazole, 99%, Thermo Scientific Chemicals Product Code.: 11304236

2-Fluorobenzothiazole, 99%, Thermo Scientific Chemicals

Product Code. 11304236
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 11304236

Brand: Thermo Scientific Alfa Aesar L19316.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1123-98-4
Molecular Formula C7H4FNS
Molecular Weight (g/mol) 153.174
MDL Number MFCD00047254
InChI Key QVWCHVAUHZEAAT-UHFFFAOYSA-N
Synonym 2-fluorobenzothiazole, 2-fluorobenzo d thiazole, 2-benzothiazolyl fluoride, benzothiazole, 2-fluoro, 2-fluoro-benzothiazole, fluorobenzothiazole, acmc-2099en, #
PubChem CID 70753
IUPAC Name 2-fluoro-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)F

Specifications

Density 1.33
Boiling Point 97°C to 98°C (16 mmHg)
Odor Characteristic
Refractive Index 1.583
Quantity 5 g
UN Number UN2810
Beilstein 971503
Formula Weight 153.18
Percent Purity 99%
Chemical Name or Material 2-Fluorobenzothiazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10244007 Certificate TypeCertificate of Analysis Date14/10/2022 Catalog NumberL19316.06,L19316.MD,L19316.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Fluorobenzothiazole, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.