Learn More
2-Imidazolidinone hemihydrate, 98+%
CAS: 121325-67-5 | C6H14N4O3 | 190.20 g/mol
233.00 DKK - 597.00 DKK
Chemical Identifiers
CAS | 121325-67-5 |
---|---|
Molecular Formula | C6H14N4O3 |
Molecular Weight (g/mol) | 190.20 |
MDL Number | MFCD02662355 |
InChI Key | KPRJGGOOWATRNT-UHFFFAOYSA-N |
Synonym | imidazolidin-2-one hydrate, 2-imidazolidinone hemihydrate, 2-imidazolidone hemihydrate, bis imidazolidin-2-one hydrate, bis imidazolidinone hydrate, 2-imidazolidinone, hydrate 2:1, imidazolidin-2-one, imidazolidin-2-one, hydrate |
PubChem CID | 2723651 |
IUPAC Name | imidazolidin-2-one;hydrate |
SMILES | O.O=C1NCCN1.O=C1NCCN1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11433248
|
Thermo Scientific Alfa Aesar
A17666.30 |
250 g |
233.00 DKK
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423248
|
Thermo Scientific Alfa Aesar
A17666.0B |
1000 g |
597.00 DKK
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Imidazolidinone hemihydrate is used as a scavenger for formaldehyde. It is utilized for removal of formaldehyde from leather, rubber and textile coating. It is also used as a raw material for the preparation of active pharmaceutical ingredients like azlocillin sodium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Imidazolidinone hemihydrate is used as a scavenger for formaldehyde. It is utilized for removal of formaldehyde from leather, rubber and textile coating. It is also used as a raw material for the preparation of active pharmaceutical ingredients like azlocillin sodium.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
Chemical Identifiers
121325-67-5 | |
190.20 | |
KPRJGGOOWATRNT-UHFFFAOYSA-N | |
2723651 | |
O.O=C1NCCN1.O=C1NCCN1 |
C6H14N4O3 | |
MFCD02662355 | |
imidazolidin-2-one hydrate, 2-imidazolidinone hemihydrate, 2-imidazolidone hemihydrate, bis imidazolidin-2-one hydrate, bis imidazolidinone hydrate, 2-imidazolidinone, hydrate 2:1, imidazolidin-2-one, imidazolidin-2-one, hydrate | |
imidazolidin-2-one;hydrate |
Specifications
121325-67-5 | |
C6H14N4O3 | |
imidazolidin-2-one hydrate, 2-imidazolidinone hemihydrate, 2-imidazolidone hemihydrate, bis imidazolidin-2-one hydrate, bis imidazolidinone hydrate, 2-imidazolidinone, hydrate 2:1, imidazolidin-2-one, imidazolidin-2-one, hydrate | |
KPRJGGOOWATRNT-UHFFFAOYSA-N | |
imidazolidin-2-one;hydrate | |
2723651 | |
≥98% |
55°C | |
MFCD02662355 | |
Soluble in water. | |
O.O=C1NCCN1.O=C1NCCN1 | |
190.20 | |
95.10 (86.09 Anhydrous) | |
2-Imidazolidinone hemihydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
H319-H361f
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.