missing translation for 'onlineSavingsMsg'
Learn More
2'-Methoxy-5'-(trifluoromethyl)acetophenone, 97%, Thermo Scientific™ Product Code.: 15445397

2'-Methoxy-5'-(trifluoromethyl)acetophenone, 97%, Thermo Scientific™

Product Code. 15445397
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15445397

Brand: Thermo Scientific Alfa Aesar H32652.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
In synthesis and evaluation of disubstituted N1- and N3-Imidazolidin-2-ones Acting as Potential Immunosuppressive Agents

Solubility
Slightly soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 503464-99-1
Molecular Formula C10H9F3O2
Molecular Weight (g/mol) 218.18
MDL Number MFCD09832300
InChI Key FPJNPSGNLVIBBS-UHFFFAOYSA-N
Synonym 1-2-methoxy-5-trifluoromethylphenyl ethanone, 1-2-methoxy-5-trifluoromethyl phenyl ethanone, 2'-methoxy-5'-trifluoromethyl acetophenone, ethanone, 1-2-methoxy-5-trifluoromethyl phenyl, 2-methoxy-5-trifluoromethyl acetophenone, 2'-methoxy-5'-trifluoromethyl-acetophenone, 1-acetyl-2-methoxy-5-trifluoromethyl benzene, 1-2-methoxy-5-trifluoromethyl phenyl ethan-1-one
PubChem CID 9942601
IUPAC Name 1-[2-methoxy-5-(trifluoromethyl)phenyl]ethanone
SMILES COC1=C(C=C(C=C1)C(F)(F)F)C(C)=O

Specifications

Melting Point 45°C to 47°C
Quantity 1 g
Solubility Information Slightly soluble.
Formula Weight 218.17
Percent Purity 97%
Chemical Name or Material 2'-Methoxy-5'-(trifluoromethyl)acetophenone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10219642 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code
Lot Number10171236 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code
Lot Number10171985 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code
Lot Number10218747 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Lot Number10153046 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2'-Methoxy-5'-(trifluoromethyl)acetophenone, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.