missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
2-Methyl-3-pentanol was used as a volatile marker. It used as a fuel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methyl-3-pentanol was used as a volatile marker. It used as a fuel.
Solubility
Amalgamate with ethanol, ethyl ether, benzene. Insoluble in water
Notes
Incompatible materials are oxidizing agents, acids, Acid chlorides, Acid anhydrides. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
2-Methyl-3-pentanol was used as a volatile marker. It used as a fuel.
Solubility
Amalgamate with ethanol, ethyl ether, benzene. Insoluble in water
Notes
Incompatible materials are oxidizing agents, acids, Acid chlorides, Acid anhydrides. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Specifications
Specifications
Density | 0.83 |
Boiling Point | 127°C to 129°C |
Flash Point | 46°C (114°F) |
Refractive Index | 1.417 |
Quantity | 10 g |
UN Number | UN2282 |
Beilstein | 1718952 |
Solubility Information | Amalgamate with ethanol,ethyl ether,benzene. Insoluble in water |
Formula Weight | 102.18 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-Methyl-3-pentanol, 97+% >
Spot an opportunity for improvement?Share a Content Correction