missing translation for 'onlineSavingsMsg'
Learn More
2-Phenylmalonamide, 97%, Thermo Scientific Chemicals Product Code.: 11461537

2-Phenylmalonamide, 97%, Thermo Scientific Chemicals

Product Code. 11461537
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11461537

Brand: Thermo Scientific Alfa Aesar B24972.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10255-95-5 | C9H10N2O2 | 178.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10255-95-5
Molecular Formula C9H10N2O2
Molecular Weight (g/mol) 178.19
MDL Number MFCD00051776
InChI Key CPSUAFUQJBJMPO-UHFFFAOYSA-N
Synonym 2-phenylmalonamide, propanediamide, 2-phenyl, phenylmethane-1,1-dicarboxamide, phenylmalonamide, 2-phenyl-malonamide, 2-phenylmalondiamide, acmc-20anni, maybridge1_000163, ksc492s5h
PubChem CID 2747536
IUPAC Name 2-phenylpropanediamide
SMILES NC(=O)C(C(N)=O)C1=CC=CC=C1

Specifications

Melting Point ∼233°C (decomposition)
Quantity 5 g
Beilstein 2446518
Formula Weight 178.19
Percent Purity 97%
Chemical Name or Material 2-Phenylmalonamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Phenylmalonamide, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.