Learn More
Fisher Chemical 200LT CHLOROFORM - SPECIFIED REAGENTAMYLENE SPECIFIED
Brand: Fisher Chemical C/4920/27
7356.51 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
200LT CHLOROFORM - SPECIFIED REAGENTAMYLENE SPECIFIED
This item is not returnable.
View return policy
Description
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Chloroform, stabilized with amylene
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H372-Causes damage to organs.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
Product Content Correction
Product Title
Fisher Chemical 200LT CHLOROFORM - SPECIFIED REAGENTAMYLENE SPECIFIED >
Spot an opportunity for improvement?Share a Content Correction