Learn More
Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh
Made from Stainless Steel and available in a wide variesty of sizes. Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh has a 200mm diameter.
Brand:  Fisherbrand™ 01/200SBW1.18
 View more versions of this product
Product Code. 11527542
  • 605.00 DKK / Each
Only null left
Estimated Shipment: 29-09-2025
Add to basket
Description
Description
100 mm and 200 mm diameter test sieves with Stainless Steel frame and woven wire mesh. Supplied with Certificate of Conformity and record card. Additional mesh sizes and diameter sieves available - please contact Customer Service for details.
Specifications
Specifications
1 Each
Sieve
Stainless Steel
200 mm
Woven Wire
1.18 mm
Certificate of Conformity, BS 410: Part 1: 2000, ISO 3310: Part 1: 2000
Certificate of conformity and record card
Stainless Steel
Certificates
View accessory products
One moment while we fetch your results.
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh > Pore Size: 1.18 mm