Learn More
Thermo Scientific 20MM Electronic Crimper
Brand: Thermo Scientific 20 00 3949
8402.04 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
20MM Electronic Crimper
This item is not returnable.
View return policy
Product Title
Thermo Scientific 20MM Electronic Crimper >
Spot an opportunity for improvement?Share a Content Correction