Learn More
uvex 2111 Magnetic Holder for One2Click Ear Plug Dispenser
Retrofit set with high adhesive force to attach to all types of magnetic surfaces. For use with Uvex One2Click Ear Plug Dispensers (2112000).
Brand: uvex 2111989
324.92 DKK valid until 2025-06-08
Use promo code "24131" to get your promotional price.
Description
- Retrofit Set for Uvex One2Click Ear Plug Dispensers
- High adhesive force attaches to all magnetic surfaces
- No drilling or screwing necessary
Specifications
One2Click Ear Plug Dispenser | |
Magnetic Holder | |
Aluminum | |
EN 352-2 |
One2Click | |
Reusable | |
Magnetic |
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08