Learn More
uvex 2112 X-Fit Disposable Ear Plugs
Ergonomic, pre-shaped ear plugs are characterized by very high insulation values making them suitable for use in extremely noisy environments.
95.15 DKK - 158.00 DKK
Specifications
Color | Lime Green |
---|---|
ANSI S3.1974 | No |
Size | Regular |
Model | 2112 X-Fit |
Noise Reduction Rating (NRR) | 37 dB |
Product Code | Brand | Packaging Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Price | Quantity & Availability | ||||||
17280342
|
uvex
2112133 |
Mini Box |
95.15 DKK
15 pairs |
In Stock
Log in to see stock. ![]() |
||||||
17240342
|
uvex
2112094 |
Polybag |
158.00 DKK
100 pairs |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
Description
- Ergonomically pre-shaped earplugs characterized by very high insulation values
- Soft foam offers a high level of comfort, even after long periods of use
- Suitable for use in extremely noisy environments
- Available in a hygienic polybag, or retail minibox
Specifications
Lime Green | |
Regular | |
37 dB | |
Oval | |
Single-use | |
No |
No | |
2112 X-Fit | |
Uncorded | |
Ear Plugs | |
No | |
EN 352-2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
uvex 2112 X-Fit Disposable Ear Plugs