Learn More
Thermo Scientific Alfa Aesar 2500ML Acetonitrile, HPLC Grade (Far UV), 99.8+%
Brand: Thermo Scientific Alfa Aesar 042880.0F
865.47 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
2500ML Acetonitrile, HPLC Grade (Far UV), 99.8+%
Safety and Handling
- Acetonitrile
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Acetonitrile
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2539095 | Certificate of Analysis | 02/06/2025 | 042880.0F,042880.K2,042880.K7 |
2538727 | Certificate of Analysis | 24/05/2025 | 042880.0F,042880.K2,042880.K7 |
2538356 | Certificate of Analysis | 24/05/2025 | 042880.0F,042880.K2,042880.K7 |
2538355 | Certificate of Analysis | 24/05/2025 | 042880.0F,042880.K2,042880.K7 |
2537429 | Certificate of Analysis | 24/05/2025 | 042880.0F,042880.K2,042880.K7 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.