missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2R,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, 98%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Specifications
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 5 g |
Solubility Information | Solubility in water: <0.1% |
Formula Weight | 263.34 |
Percent Purity | 98% |
Chemical Name or Material | (2R, 3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane |
Safety and Handling
Product Identifier
- (2R,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
(2R,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, 98%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction