Learn More
DWK Life Sciences Kimble Chase™ 3.3 Borosilicate Glass Test Tubes
Use round-bottom 3.3 Borosilicate Glass Test Tubes for laboratory needs when heat resistance is critical. These disposable test tubes are thick-walled and autoclavable.
Brand: DWK Life Sciences A613014000911
233.70 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
- Disposable
- Borosilicate glass
- Excellent resistance to temperature changes
- Autoclavable
Specifications
Round bottom | |
100Pack | |
Test Tube |
130 mm | |
14 mm |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
67Hours
19Minutes
40Seconds
20New Offer Save Up to 85%