Learn More
3,4-Dichloro-1,2,5-thiadiazole, 99%, Thermo Scientific Chemicals
Description
3,4-Dichloro-1,2,5-thiadiazole is a chlorinated thiadiazole that is a potent inhibitors of nitrification in soil. 3,4-Dichloro-1,2,5-thiadiazole is used as a building block in the preparaton of the β-adrenergic blocking agent, Timolol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3,4-Dichloro-1,2,5-thiadiazole is a chlorinated thiadiazole that is a potent inhibitors of nitrification in soil. 3,4-Dichloro-1,2,5-thiadiazole is used as a building block in the preparaton of the β-adrenergic blocking agent, Timolol.
Solubility
Soluble in chloroform and methanol.
Notes
Hygroscopic. Store under dry inert gas. Incompatible with moisture and oxidizing agents, acids and bases.

Specifications
Specifications
Density | 1.644 |
Boiling Point | 157°C to 159°C |
Flash Point | >100°C (212°F) |
Refractive Index | 1.562 |
Quantity | 10 g |
UN Number | UN2810 |
Beilstein | 606585 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in chloroform and methanol. |
Formula Weight | 155.01 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10243865 | Certificate of Analysis | 13/10/2022 | A11738.09,A11738.18,A11738.30 |
5002P17P | Certificate of Analysis | 22/11/2021 | A11738.09,A11738.18,A11738.30 |
10106170 | Certificate of Analysis | 22/11/2021 | A11738.09,A11738.18,A11738.30 |
10205798 | Certificate of Analysis | 22/11/2021 | A11738.09,A11738.18,A11738.30 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.