missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-3-heptanol, erythro + threo, 98+%, Thermo Scientific™

Product Code. 11409936
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in the synthesis of tertiary 14C-labelled nonylphenol isomers..

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19549-74-7
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.26
MDL Number MFCD00046585
InChI Key NOSOEGQGQOMHBF-UHFFFAOYNA-N
Synonym 3,5-dimethyl-3-heptanol, 3-heptanol, 3,5-dimethyl, 3,5-dimethyl-3-heptanol, erythro + threo, acmc-20ance, 4-heptanol,3,5-dimethyl, ethyl methyl 2-methylbutyl carbinol
PubChem CID 140546
IUPAC Name 3,5-dimethylheptan-3-ol
SMILES CCC(C)CC(C)(O)CC

Specifications

Quantity 50 g
Formula Weight 144.25
Percent Purity ≥98%
Chemical Name or Material 3,5-Dimethyl-3-heptanol, erythro + threo
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'