missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethyl-3-heptanol, erythro + threo, 98+%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used in the synthesis of tertiary 14C-labelled nonylphenol isomers..
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used in the synthesis of tertiary 14C-labelled nonylphenol isomers..
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
Quantity | 10 g |
Formula Weight | 144.25 |
Percent Purity | ≥98% |
Chemical Name or Material | 3,5-Dimethyl-3-heptanol, erythro + threo |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'