missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dimethyl-3-heptanol, 99%, Thermo Scientific™

Product Code. 11409796
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1573-28-0
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.26
MDL Number MFCD00021828
InChI Key HVPGGLNDHUWMLS-UHFFFAOYNA-N
PubChem CID 15297
IUPAC Name 3,6-dimethylheptan-3-ol
SMILES CCC(C)(O)CCC(C)C

Specifications

Density 0.82
Quantity 50 g
Synonym 3,6-dimethyl-3-heptanol, 3-heptanol, 3,6-dimethyl, acmc-20anbp, 3-heptanol,3,6-dimethyl
Formula Weight 144.25
Percent Purity 99%
Chemical Name or Material 3,6-Dimethyl-3-heptanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.