missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4,4-dimethylpentanoic acid hydrate, 98%, Thermo Scientific™

Product Code. 10235481
Click to view available options
Quantity:
500 mg
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Melting Point 223°C to 225°C
CAS 336185-29-6
CAS Min % 97.5
CAS Max % 100.0
Color White
Assay Percent Range 98%
Molecular Formula C7H15NO2
Infrared Spectrum Authentic
Linear Formula (H3C)3CCH(NH2)CH2COOH
MDL Number MFCD03791052
Packaging Glass Bottle
Synonym 3-amino-4,4-dimethylpentanoic acid xhydrate, 3-amino-4,4-dimethylpentanoic acid hydrate, 3-amino-4,4-dimethylpentanoic acid-water 1/1
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol
InChI Key MIMSUZKTGRXZNZ-UHFFFAOYNA-N
SMILES CC(C)(C)C(N)CC(O)=O
IUPAC Name 3-amino-4,4-dimethylpentanoic acid;hydrate
Molecular Weight (g/mol) 145.20
PubChem CID 53393206
Formula Weight 145.2
Percent Purity 98%
Quantity 500 mg
Physical Form Crystalline Flakes
Chemical Name or Material 3-Amino-4, 4-dimethylpentanoic acid hydrate, 98%
Show More Show Less
Product Identifier
  • 3-Amino-4,4-dimethylpentanoic acid hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'