Learn More
3'-Azido-3'-deoxy-5-fluorocytidine, MedChemExpress
MedChemExpress 3'-Azido-3'-deoxy-5-fluorocytidine (Compound 12) is a cytidine derivative.
Brand:  Medchem Express HY-111641
Encompass_Preferred
 View more versions of this product
Product Code. 16434508
  • 1489.66 DKK / 10mg
Explore more special offers
Description
Description
3'-Azido-3'-deoxy-5-fluorocytidine (Compound 12) is a cytidine derivative.
Specifications
Specifications
3'-Azido-3'-deoxy-5-fluorocytidine
White
C9H11FN6O4
DMSO : 140 mg/mL (489.13 mM; Need ultrasonic)
286.22
99.98%
Solid
2095417-18-6
Research
10 mg
O=C1N([C@@H]2O[C@H](CO)[C@@H](N=[N+]=[N-])[C@H]2O)C=C(F)C(N)=N1
286.22
Research
Neuroscience-Neuromodulation
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.