Learn More
3'-Azido-3'-deoxy-5-fluorocytidine, MedChemExpress
MedChemExpress 3'-Azido-3'-deoxy-5-fluorocytidine (Compound 12) is a cytidine derivative.
Brand: Medchem Express HY-111641
This item is not returnable.
View return policy
Description
3'-Azido-3'-deoxy-5-fluorocytidine (Compound 12) is a cytidine derivative.Specifications
3'-Azido-3'-deoxy-5-fluorocytidine | |
White | |
C9H11FN6O4 | |
DMSO : 140 mg/mL (489.13 mM; Need ultrasonic) | |
286.22 | |
99.98% | |
Solid |
2095417-18-6 | |
Research | |
10 mg | |
O=C1N([C@@H]2O[C@H](CO)[C@@H](N=[N+]=[N-])[C@H]2O)C=C(F)C(N)=N1 | |
286.22 | |
Research | |
Neuroscience-Neuromodulation |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.