missing translation for 'onlineSavingsMsg'
Learn More
3-Bromo-1-propanol, 95%, Thermo Scientific Chemicals Product Code.: 11429897

3-Bromo-1-propanol, 95%, Thermo Scientific Chemicals

Product Code. 11429897
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11429897

Brand: Thermo Scientific Alfa Aesar B22857.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Questo articolo non è restituibile. Consulta la politica di reso

3-Bromo-1-propanol is used in the synthesis of fluorescent halide-sensitive quinolinium dyes, chiral, quaternary prolines through cyclization of quaternary amino acids and molten salt-polymers. It is utilized for the study of micellar media and in microemulsions based on cationic or a nonionic surfactant by reacting with phenols. It acts as a grafting agent in the synthesis of recyclable reagents and an intermediate in the synthesis of organic molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-1-propanol is used in the synthesis of fluorescent halide-sensitive quinolinium dyes, chiral, quaternary prolines through cyclization of quaternary amino acids and molten salt-polymers. It is utilized for the study of micellar media and in microemulsions based on cationic or a nonionic surfactant by reacting with phenols. It acts as a grafting agent in the synthesis of recyclable reagents and an intermediate in the synthesis of organic molecules.

Solubility
Miscible with water. Slightly miscible with ethanol and ether.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 627-18-9
Molecular Formula C3H7BrO
Molecular Weight (g/mol) 138.99
MDL Number MFCD00002942
InChI Key RQFUZUMFPRMVDX-UHFFFAOYSA-N
Synonym 3-bromo-1-propanol, trimethylene bromohydrin, 1-propanol, 3-bromo, 3-bromopropanol, 3-hydroxypropyl bromide, 1-bromo-3-propanol, 3-bromo-propan-1-ol, ccris 5979, 1-bromo-3-hydroxypropane, unii-3t4axh68fhShow MoreShow Less
PubChem CID 12308
IUPAC Name 3-bromopropan-1-ol
SMILES OCCCBr

Specifica

Density 1.59
Boiling Point 71°C to 73°C (11 mmHg)
Flash Point 93°C (199°F)
Linear Formula BrCH2CH2CH2OH
Refractive Index 1.4885
Quantity 25 g
Beilstein 969160
Solubility Information Miscible with water. Slightly miscible with ethanol and ether.
Formula Weight 138.99
Percent Purity ≥97%
Chemical Name or Material 3-Bromo-1-propanol
Vedi altri risultati Mostra meno risultati

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10246038 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberB22857.14,B22857.06,B22857.22
Lot Number10152451 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22857.14,B22857.06,B22857.22
Lot Number10166146 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22857.14,B22857.06,B22857.22
Lot Number10235875 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22857.14,B22857.06,B22857.22
Lot Number10182558 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22857.14,B22857.06,B22857.22
compliance-icons
Identificativo prodotto
  • 3-Bromo-1-propanol
Avvertenza
  • Warning
Categoria di pericolo
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Consigli di prudenza
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Informazioni supplementari
  • MIXTURE LIST-Contains : 3-Bromo-1-propanol

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
3-Bromo-1-propanol, 95%, Thermo Scientific Chemicals >

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato