missing translation for 'onlineSavingsMsg'
Learn More
3-Bromo-2,4,5-trifluorobenzoic acid, 98% Product Code.: 11398956

3-Bromo-2,4,5-trifluorobenzoic acid, 98%

Product Code. 11398956
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11398956

Brand: Thermo Scientific Alfa Aesar L20154.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 104222-42-6 | C7H2BrF3O2 | 254.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104222-42-6
Molecular Formula C7H2BrF3O2
Molecular Weight (g/mol) 254.99
MDL Number MFCD05664065
InChI Key VWVLSESGZBAPHA-UHFFFAOYSA-N
Synonym 2,4,5-trifluoro-3-bromobenzoic acid, benzoic acid, 3-bromo-2,4,5-trifluoro, 3-bromanyl-2,4,5-tris fluoranyl benzoic acid, pubchem1366, 3-bromo-2,4,5-trifluoro-benzoic acid, acmc-20an96, 3-bromo-2,4,5-trifluoro benzoic acidShow MoreShow Less
PubChem CID 2783246
IUPAC Name 3-bromo-2,4,5-trifluorobenzoic acid
SMILES C1=C(C(=C(C(=C1F)F)Br)F)C(=O)O

Specifications

Melting Point 122°C to 123°C
Quantity 1 g
Beilstein 7584381
Formula Weight 254.99
Percent Purity 98%
Chemical Name or Material 3-Bromo-2,4,5-trifluorobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromo-2,4,5-trifluorobenzoic acid, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.