missing translation for 'onlineSavingsMsg'
Learn More
3-(Bromomethyl)-5-methylisoxazole, 97%, Thermo Scientific™ Product Code.: 10542472

3-(Bromomethyl)-5-methylisoxazole, 97%, Thermo Scientific™

Product Code. 10542472
1 g, Amber Glass Bottle, 1g
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10542472

Brand: Thermo Scientific Maybridge CC10408DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 130628-75-0
Molecular Formula C5H6BrNO
Molecular Weight (g/mol) 176.01
MDL Number MFCD02677720
InChI Key ASGJFGPILHALRC-UHFFFAOYSA-N
Synonym 3-bromomethyl-5-methylisoxazole, 3-bromomethyl-5-methyl-1,2-oxazole, 3-bromomethyl-5-methyl-isoxazole, isoxazole,3-bromomethyl-5-methyl, zlchem 1313, acmc-1ca86, 3-bromomethyl-5-methylisoxazol, 5-brimomethyl-3-methylisoxazle
PubChem CID 2776304
IUPAC Name 3-(bromomethyl)-5-methyl-1,2-oxazole
SMILES CC1=CC(CBr)=NO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 176.01
Percent Purity 97%
Chemical Name or Material 3-(Bromomethyl)-5-methylisoxazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(Bromomethyl)-5-methylisoxazole, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.