missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(trifluoromethylthio)aniline, 97%, Thermo Scientific Chemicals

Product Code. 11465507
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64628-74-6
Molecular Formula C7H5ClF3NS
Molecular Weight (g/mol) 227.63
MDL Number MFCD00190131
InChI Key BLOPJEJSRKMVSU-UHFFFAOYSA-N
Synonym 3-chloro-4-trifluoromethylthio aniline, 3-chloro-4-trifluoromethyl thio aniline, 3-chloro-4-trifluoromethyl sulfanyl aniline, benzenamine,3-chloro-4-trifluoromethyl thio, 3-chloro-4-trifluoromethylthio phenylamine, pubchem2950, 3-chloro-4-trifluoromethylthioaniline, 3-chloro-4-trifluoromethylsulfanyl aniline, 3-chloranyl-4-trifluoromethylsulfanyl aniline, 3-chloro-4-trifluoromethylsulfanyl-phenylamine
PubChem CID 2736713
IUPAC Name 3-chloro-4-(trifluoromethylsulfanyl)aniline
SMILES NC1=CC=C(SC(F)(F)F)C(Cl)=C1

Specifications

Melting Point 41°C to 44°C
Quantity 1 g
UN Number UN2811
Beilstein 6314200
Formula Weight 227.64
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-(trifluoromethylthio)aniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'