Learn More
(3-Cyanopropyl)triethoxysilane, 97%, Thermo Scientific Chemicals
401.16 DKK - 1184.72 DKK
Chemical Identifiers
CAS | 1067-47-6 |
---|---|
Molecular Formula | C10H21NO3Si |
Molecular Weight (g/mol) | 231.367 |
MDL Number | MFCD00042636 |
InChI Key | VGIURMCNTDVGJM-UHFFFAOYSA-N |
Synonym | 3-cyanopropyltriethoxysilane, butanenitrile, 4-triethoxysilyl, 4-triethoxysilyl butanenitrile, 4-triethoxysilyl butyronitrile, butyronitrile, 4-triethoxysilyl, silane, triethoxy 3-cyanopropyl, triethoxy-3-kyanpropylsilan, unii-n78w6a24dx, 3-cyanopropyl triethoxysilane, triethoxy-3-kyanpropylsilan czech |
PubChem CID | 14024 |
IUPAC Name | 4-triethoxysilylbutanenitrile |
SMILES | CCO[Si](CCCC#N)(OCC)OCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11317425
|
Thermo Scientific Alfa Aesar
L05367.14 |
25 g |
401.16 DKK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327425
|
Thermo Scientific Alfa Aesar
L05367.22 |
100 g |
1184.72 DKK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(3-Cyanopropyl)triethoxysilane is used as cyanoalkyl silane coupling agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(3-Cyanopropyl)triethoxysilane is used as cyanoalkyl silane coupling agent.
Solubility
Not miscible in water.
Notes
Moisture Sensitive. Incompatible materials are Oxidizing agents and water.
Chemical Identifiers
1067-47-6 | |
231.367 | |
VGIURMCNTDVGJM-UHFFFAOYSA-N | |
14024 | |
CCO[Si](CCCC#N)(OCC)OCC |
C10H21NO3Si | |
MFCD00042636 | |
3-cyanopropyltriethoxysilane, butanenitrile, 4-triethoxysilyl, 4-triethoxysilyl butanenitrile, 4-triethoxysilyl butyronitrile, butyronitrile, 4-triethoxysilyl, silane, triethoxy 3-cyanopropyl, triethoxy-3-kyanpropylsilan, unii-n78w6a24dx, 3-cyanopropyl triethoxysilane, triethoxy-3-kyanpropylsilan czech | |
4-triethoxysilylbutanenitrile |
Specifications
238°C to 241°C | |
C10H21NO3Si | |
MFCD00042636 | |
74°C (165°F) | |
Not miscible in water. | |
CCO[Si](CCCC#N)(OCC)OCC | |
231.367 | |
231.37 | |
1.417 | |
0.961 |
1067-47-6 | |
NCCH2CH2CH2Si(OCH2CH3)3 | |
1942237 | |
3-cyanopropyltriethoxysilane, butanenitrile, 4-triethoxysilyl, 4-triethoxysilyl butanenitrile, 4-triethoxysilyl butyronitrile, butyronitrile, 4-triethoxysilyl, silane, triethoxy 3-cyanopropyl, triethoxy-3-kyanpropylsilan, unii-n78w6a24dx, 3-cyanopropyl triethoxysilane, triethoxy-3-kyanpropylsilan czech | |
VGIURMCNTDVGJM-UHFFFAOYSA-N | |
4-triethoxysilylbutanenitrile | |
14024 | |
97% | |
Moisture sensitive | |
(3-Cyanopropyl)triethoxysilane |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
missing translation for 'einecsNumber' : 213-931-4
missing translation for 'rtecsNumber' : VV2785000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.