missing translation for 'onlineSavingsMsg'
Learn More
3-Dimethylamino-1-(4-fluorophenyl)-2-propen-1-one, 97%, Thermo Scientific™ Product Code.: 15458745

3-Dimethylamino-1-(4-fluorophenyl)-2-propen-1-one, 97%, Thermo Scientific™

Product Code. 15458745
5 g, 5g
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15458745

Brand: Thermo Scientific Alfa Aesar H34021.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75175-77-8
Molecular Formula C11H12FNO
Molecular Weight (g/mol) 193.22
MDL Number MFCD00111509
InChI Key GTJNUCRUOWBWEW-UHFFFAOYSA-N
Synonym 2e-3-dimethylamino-1-4-fluorophenyl prop-2-en-1-one, e-3-dimethylamino-1-4-fluorophenyl prop-2-en-1-one, 3-dimethylamino-1-4-fluorophenyl-2-propen-1-one, e-3-dimethylamino-1-4-fluoro-phenyl-propenone, 4-fluorophenyl 2-dimethylamino ethenyl ketone, e-4'-fluoro-beta-dimethylamino acrylophenone, propenone, 1-4-fluorophenyl-3-dimethylamino, 2-propen-1-one, 1-4-fluorophenyl-3-dimethylamino, 2e-3-dimethylamino-1-4-fluorophenyl-2-propen-1-one #Show MoreShow Less
PubChem CID 5374477
IUPAC Name (E)-3-(dimethylamino)-1-(4-fluorophenyl)prop-2-en-1-one
SMILES CN(C)C=CC(=O)C1=CC=C(F)C=C1

Specifications

Melting Point 97°C to 99°C
Quantity 5 g
Formula Weight 193.22
Percent Purity 97%
Chemical Name or Material 3-Dimethylamino-1-(4-fluorophenyl)-2-propen-1-one

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Product Code
Lot Number10169961 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Dimethylamino-1-(4-fluorophenyl)-2-propen-1-one, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.