missing translation for 'onlineSavingsMsg'
Learn More
3-Ethoxyacrylic acid, 95%, Thermo Scientific™ Product Code.: 10289822

3-Ethoxyacrylic acid, 95%, Thermo Scientific™

Product Code. 10289822
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10289822

Brand: Thermo Scientific Maybridge RH00924DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6192-01-4
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
InChI Key SYMAGJYJMLUEQE-ONEGZZNKSA-N
Synonym 3-ethoxyacrylic acid, e-3-ethoxyacrylic acid, 3-ethoxyprop-2-enoic acid, 3-ethyoxyacrylic acid, 3-ethoxy-2-propenoic acid, 2e-3-ethoxyprop-2-enoic acid, 2e-3-ethoxyacrylic acid, 3-ethoxyacrylicacid, pubchem20958, e-3-ethoxy-acrylic acid
PubChem CID 5709609

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 116.12
Percent Purity 95%
Chemical Name or Material 3-Ethoxyacrylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Ethoxyacrylic acid, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.