Learn More
Fisherbrand™ Three-Way Swivel Scaffolding Connectors
Used for fixing three rods or tubes mutually at right angles. Fisherbrand™ Three-Way Swivel Scaffolding Connectors can be tightened by hand.
Brand:  Fisherbrand™ 2115
Product Code. 11548112
  • 109.00 DKK / Each
Only null left
Estimated Shipment: 29-08-2025
Add to basket
Description
Description
  • With thumbscrews
  • For rods and tubes of 6 to 13mm diameters
Specifications
Specifications
0 to 13 mm
3 Rods Bossehead 0-13mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Three-Way Swivel Scaffolding Connectors > Outer Diameter: 0 to 13mm