Learn More
Cole-Parmer™ 304 Stainless Steel Cover
For use with stainless steel beakers
Brand:  Cole-Parmer™ 07207-80
 View more versions of this product
Product Code. 11797974
  • 431.00 DKK / Each
Only null left
Estimated Shipment: 26-09-2025
Description
  • Covers reduce evaporation
  • Contain harmful vapors, or keep contaminants out
Specifications
Specifications
222mm
Stainless Steel (304)
1Each
AISI 304
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cole-Parmer™ 304 Stainless Steel Cover > Diameter: 600mm